Learn More
Nelson Jameson Company MINI FLPTOP 10ML DE BRTH 100CS
Supplier:  Nelson Jameson Company 3513310 C
Encompass
This item has a minimum order quantity of 4 per supplier requirements. "•Hinged lid can be opened with one hand and stays open for easy sample additions. •Decontaminated through gamma irradiation and ready for use with no additional blending required. •Flat bottom tube stands on its own, no holder required. •Leak resistant vial helps prevent leakage during shipping.
Catalog No. NC1115208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.