Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mini Gel Tank Lid

Numéro de catalogue. A25944
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A25944 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A25944 Fournisseur Invitrogen™ Code fournisseur A25944
  • $224.00
    $217.65 / Each of 1

    Save $6.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Afficher toutes les promotions
Ajouter au panier
Edge
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Mini Gel Tank Lid is a replacement part for the Mini Gel Tank. It goes on top of the Gel Runner Tank.

About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Spécifications

For Use With (Equipment) Mini Gels
Gel Compatibility Novex™ Mini Gels
Gel Size Mini
Type Mini Gel Tank Lid
Quantity 1 unit
Shipping Condition Room Temperature
Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

What are the storage conditions for Bolt gels?

Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.

Can I purchase replacement parts for the Mini Gel Tank?

Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732

What are the components of the Mini Gel Tank?

The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.