Learn More
Wards Natural Science Est Ward's® Mini Magnifier, 1.75×, 16.1×13.5×20.7 cm
Supplier:  Wards Natural Science Est 470311198
Encompass
The Ward's® mini magnifier is mounted on a stand to keep hands free for manipulating samples. Excellent for viewing bacterial colonies on media, the magnifier pairs well with the Ward's® mini light box and Ward's®
Catalog No. NC2878269