Learn More
Key Scientific Products Mini Slide Warmer, 23 Slide Capacity, LED Temperature Display
SDP
Catalog No. NC0987506
Encompass_Preferred
Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting and slide fixation prior to staining. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display. 10" X 8", 23 slide capacity, Temp range to 70 C.
Catalog No. NC0987506 Supplier Key Scientific Products Supplier No. CY21297
Ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting and slide fixation prior to staining. Thermal heater ensures even heat transfer. Anodized black surface provides contrast. LED temperature display. 10" X 8", 23 slide capacity, Temp range to 70 C.