Learn More
BURKLE INC PTFE-valve
Supplier:  BURKLE INC 86100006
Encompass_Preferred
Three-way valves made of solid PTFE with hose nozzles Resistant to chemicals Very easy connection thanks to its excellent antifriction properties Blue handles made of PP
Catalog No. 50-000-25720
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.