Learn More
Thomas Scientific Mini Ultrasonic Cleaner
Supplier: Thomas Scientific 1226Z49
Ultrasonic Cleaners are designed to remove grease, dust or other contaminants from surfaces & cavities of small objects, parts & samples such as cuvettes, lenses, glassware, stainless steel instruments, metals & more. Simply place objects directly into the stainless steel tank or into a dipping basket, replace the transparent lid & press "on". The transducer generates thousands of minute cleansing bubbles which vibrate at a very high electrical frequency. When the vibration speed rises above the ultrasonic frequency level, bubbles explode & generate strong power, cleaning the surfaces & cavities of hard-to-clean objects. Ultrasonic cleaning does not scratch the surface or harm objects like many chemical or abrasive cleansers. Units operate quietly, powered by AC 100/120V. High Power Ultrasonic Cleaner 100004 includes a dipping basket & features 5 pre-programmed timing cycles with the remaining count-down time shown on a large bright LED. A cooling fan enables prolonged op
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.