Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Mini Weather Station
SDPSCI_ED

Catalog No. S111030 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S111030 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111030 Supplier United Scientific™ Supplier No. MWS136
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Introduce basic weather concepts with this versatile apparatus.

  • Measure temperature (in Fahrenheit and Celsius) and wind speed, then reference the screen-printed chart to easily calculate wind chill.
  • The device freely rotates to indicate wind direction, and its removable rain gauge allows for the quick assessment of rainfall.
  • The product is designed to mount on posts.

Specifications

Grade 6 to 12
Format Kit
Product Type Weather Station
Languages English
Subject Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.