Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Miniature DC Motor
SDPSCI_ED

Catalog No. S15693 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
DC Motor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S15693 DC Motor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15693 Supplier United Scientific™ Supplier No. DCM015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General purpose motor is an Ideal choice for many electricity demonstrations.

  • Operates on 1.5 V or 3 V DC.
  • Speed: 7000 to 11000 RPM.
  • Motor is approximately 40 mm long x 20 mm in diameter
  • Shaft size is 8 mm long x 2 mm in diameter
  • Includes soldered-on on 4 in. wire leads

Specifications

For Use With (Application) Electrical Experiments
Product Type DC Motor

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.