Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva MiniBeads™ Columns

Catalog No. 45000307 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Mini Q Ion Exchange Column
Bed Volume:
0.8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Bed Volume
45-000-307 Mini Q Ion Exchange Column 0.8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-000-307 Supplier Cytiva Supplier No. 17517701/DEL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MiniBeads™ prepacked columns are designed for high performance purification and characterization of biomolecules using ion exchange chromatography

  • Small, mono-disperse beads for extremely high resolution purifications. non porous beads contribute to high resolution but give lower capacity than MonoBeads
  • Analytical and micro-preparative applications with choice of both anion and cation exchangers
  • High chemical and physical stability
  • Excellent reproducibility and durability

  • Tricorn high-performance columns (4.6/50 PE) prepacked with MiniBeads
  • PC columns designed for micropurification

Precision column holders may be required for use on high performance chromatography systems such as KTA design. For optimum results, it is important to check the compatibility of the column and the system specifications.

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
Product Type Mini Q Ion Exchange Column
Bed Volume 0.8 mL
Diameter (English) Inner 0.18 in.
Diameter (Metric) Inner 4.6 mm
Quantity Each
Height (English) 1.96 in.
Height (Metric) 50 mm
Flow Rate 0.5 to 2 mL/min.
Includes Valco fittings
Material Polyethere Ketone
Ligand Type O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3
Max. Pressure 180psi
pH Range 3 to 11
Particle Size 3 μm
Support Type Monodisperse Polystyrene/Divinyl Benzene particles
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) High performance purification and characterization of biomolecules by ion exchange chromato graph
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.