Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-GARD™ Specimen Biohazard Transport Bags

Catalog No. 22307935
Change view
Click to view available options
Dimensions (L x W):
6 x 6 in.
9 x 5 in.
9 x 6 in.
10 x 6 in.
10 x 8 in.
10 x 10 in.
17 x 15 in.
18 x 13 in.
Includes:
1 Pocket
1 Pocket, Space for indicating storage method
2 Pockets
2 Pockets, 3 x 3 in. Absorbent Sheet
2 Pockets, Instructions
2 Pockets, Red and Black Print
Instructions
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Includes
22-307935 9 x 5 in. 2 Pockets
22-026754 18 x 13 in. 1 Pocket, Space for indicating storage method
22-314257 10 x 6 in. 2 Pockets
22-026753 17 x 15 in. 2 Pockets
22-307-948 10 x 10 in. 2 Pockets, Instructions
22-307945 10 x 8 in. 2 Pockets
22-310-094 6 x 6 in. 1 Pocket
22-310-001 10 x 6 in. 2 Pockets, Red and Black Print
22-310-004 10 x 6 in. Instructions
22-310-095 9 x 6 in. 2 Pockets, 3 x 3 in. Absorbent Sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 22-307935 Supplier Minigrip™ Supplier No. 95CMS59
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for specimen transport in hospitals and labs

  • Features liquid-tight, Press and Close™ adhesive closure system — simply peel away the release liner and press to close
  • With printed biohazard logo
  • Separate outside pocket holds the requisition slip
  • Traditional scored opening
  • No glove-to-adhesive contact, ensuring ease of use
  • Once closed, the bag stays closed, even in pneumatic tube systems
  • Remains closed if air is trapped inside, or if “grabbed,” will not “pop open”
  • Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging
  • To open, simply snap at score
  • Disposable, eliminating the possibility of cross-contamination caused by reusing bags
  • Manufactured with Micro-Tex™ polyethylene for slip-free handling

Order Info

Cat. No. 22-310-001 contains an additional pre-inserted super absorbent tested to absorb up to 60mL of liquid.

Specifications

Closure Type Press and Close Liquid Tight
Color Clear
Disposable Single-use
For Use With (Application) Used for specimen transport in hospitals and labs
Includes 2 Pockets
Material Polyethylene
Dimensions (L x W) 9 x 5 in.
Length (English) 9 in.
Width (English) 5 in.
Height (English) 9 in.
Height (Metric) 22.86 cm
Product Type Biohazard Specimen Transport Bag
Length (Metric) 22.86 cm
Width (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.