Learn More
Minigrip™ SPECI-GARD™ Specimen Biohazard Transport Bags
Description
- Features liquid-tight, Press and Close™ adhesive closure system — simply peel away the release liner and press to close
- With printed biohazard logo
- Separate outside pocket holds the requisition slip
- Traditional scored opening
- No glove-to-adhesive contact, ensuring ease of use
- Once closed, the bag stays closed, even in pneumatic tube systems
- Remains closed if air is trapped inside, or if “grabbed,” will not “pop open”
- Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging
- To open, simply snap at score
- Disposable, eliminating the possibility of cross-contamination caused by reusing bags
- Manufactured with Micro-Tex™ polyethylene for slip-free handling
Order Info
Cat. No. 22-310-001 contains an additional pre-inserted super absorbent tested to absorb up to 60mL of liquid.
Specifications
Specifications
| Closure Type | Press and Close Liquid Tight |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Application) | Used for specimen transport in hospitals and labs |
| Includes | 2 Pockets, Instructions |
| Material | Polyethylene |
| Dimensions (L x W) | 10 x 10 in. |
| Length (English) | 10 in. |
| Width (English) | 8 in. |
| Height (English) | 10 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.