Learn More
Minigrip™ SPECI-ZIP™ Reclosable Colored Biohazard Bags
Description
- Colored or clear transparent plastic bags with biohazard logo
- Bilingual in English and Spanish
Order Info
Case of 1000 in packs of 100.
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Designed with info pocket, tear line |
| Includes | Printed biohazard warning, info pocket, tear line |
| Width (English) | 6 in. |
| Height (English) | 9 in. |
| Height (Metric) | 22.86 cm |
| Product Type | Reclosable Biohazard Bag |
| Width (Metric) | 15.24 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.