Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-ZIP™ Reclosable Colored Biohazard Bags

Catalog No. 22310413
Change view
Click to view available options
Color:
Blue
Clear
Green
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-310-413 Green
22-310-407 Clear
22-310-412 Yellow
22-310-411 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-310-413 Supplier Minigrip™ Supplier No. IP69BG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed with info pocket (three wall) and tear line

  • Colored or clear transparent plastic bags with biohazard logo
  • Bilingual in English and Spanish

Order Info

Case of 1000 in packs of 100.

Specifications

Color Green
For Use With (Application) Designed with info pocket, tear line
Includes Printed biohazard warning, info pocket, tear line
Width (English) 6 in.
Height (English) 9 in.
Height (Metric) 22.86 cm
Product Type Reclosable Biohazard Bag
Width (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.