Learn More
REPLIGEN CORPORATION MINIKROS SAMPLER 300 KDA 0.5 M
SDP
Supplier:  REPLIGEN CORPORATION S04E30005S
Encompass
Minikros Sampler 300 kDa 0.5 mm fiber, mPES 1600 cm2 surface area hollow fiber
Catalog No. NC2108489
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.