Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MiniMark™ Industrial Label Printer Ribbons

Catalog No. 19104550 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Red
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-550 Blue
19-104-547 Black
19-104-548 White
19-104-552 Yellow
19-104-549 Red
19-104-551 Green
19-104-553 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-104-550 Supplier Brady™ Supplier No. 103028

Create affordable custom identification with the MiniMark industrial label printer. Brady™ MiniMark™ Industrial Label Printer Ribbons are available in a variety of colors.

  • Pre-loaded onto spindles with'keyed'end caps
  • Tough, smear proof ribbons produce long-lasting printed text
  • For industrial and outdoor areas

Specifications

Product Type Printer Ribbon
Color Blue
For Use With (Equipment) MiniMark Printer
Length (English) 290 ft.
Dimensions (L x W) 290 ft. x 4 in.
Quantity 2/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.