Learn More
Cytiva Cytiva Minimate EVO Reservoir Kit, High Protein Recovery, Versatile Lab Applications, Unit Each
Catalog No. NC3310524 Shop All Cytiva Products
Encompass
The Cytiva Minimate EVO Reservoir Kit is crucial for the Minimate EVO Tangential Flow Filtration system, enhancing bioprocess efficiency in biochemistry. It aids in concentrating, desalting, buffer exchanging, and fractionating biological samples, ensuring high sample recovery rates while minimizing contamination. Constructed with biocompatible materials like polysulfone and polypropylene, it supports both new setups and upgrades of older systems. This adaptability and reliability make it stand out for laboratory-scale applications.
Catalog No. NC3310524 Supplier Cytiva Supplier No. 97008
The Cytiva Minimate EVO Reservoir Kit is crucial for the Minimate EVO Tangential Flow Filtration system, enhancing bioprocess efficiency in biochemistry. It aids in concentrating, desalting, buffer exchanging, and fractionating biological samples, ensuring high sample recovery rates while minimizing contamination. Constructed with biocompatible materials like polysulfone and polypropylene, it supports both new setups and upgrades of older systems. This adaptability and reliability make it stand out for laboratory-scale applications.