Learn More
AMPLYUS LLC miniPCR mini16 thermal cycler
Supplier: AMPLYUS LLC QP101601
Small and portable, the mini16 thermal cycler fits right on your bench and travels wherever your science goes. The mini16 offers 16-well capacity and Bluetooth connectivity with computers and mobile devices.The ultimate miniPCR machine. Proven performance. Ideal for lab, classroom, and field use. Weighs under one pound, fits in the palm of your hand. AC or battery powered (battery not included).Easy to use via intuitive software. PCR of choice aboard the International Space Station.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.