Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 131371AP150UL
Description
This gene encodes a serine/threonine kinase belonging to the germinal center kinase (GCK) family. The protein is structurally similar to the kinases that are related to NIK and may belong to a distinct subfamily of NIK-related kinases within the GCK family. Studies of the mouse homolog indicate an up-regulation of expression in the course of postnatal mouse cerebral development and activation of the cJun N-terminal kinase (JNK) and the p38 pathways.Specifications
| MINK1 | |
| Polyclonal | |
| Unconjugated | |
| MINK1 | |
| B55, GCK family kinase MiNK, hMINK, hMINKbeta, MAP4K6, MEK kinase kinase 6, MEKKK 6, MINK, MINK1, Misshapen like kinase 1, Misshapen/NIK related kinase, YSK2, ZC3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303259, 50488, 50932 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F1LP90, Q8N4C8, Q9JM52 | |
| Mink1 | |
| MINK1 Fusion Protein Ag3795 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction