Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minocycline HCl, USP Grade, G-Biosciences
SCI_ED

Catalog No. S157214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157214 5 g
S157215 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157214 Supplier G-Biosciences Supplier No. RC1054
Add to Cart
Add to Cart

Minocycline HCl is a broad-spectrum tetracycline antibiotic.

It has been shown to inhibit endothelial cell proliferation and angiogenesis and the replication of HIV. It has also displayed anti-inflammatory and anti-collagenase activity.

Chemical Identifiers

CAS 13614-98-7
Molecular Weight (g/mol) 493.94 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 5 g
Percent Purity 98%
Grade USP
Chemical Name or Material Minocycline HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.