CAS RN 13614-98-7
Thermo Scientific Chemicals Minocycline hydrochloride, 97%
CAS: 13614-98-7 Molecular Formula: C23H28ClN3O7 Molecular Weight (g/mol): 493.94 MDL Number: MFCD00083669 InChI Key: KDLQIOPKJDNQIM-WUURTAMISA-N PubChem CID: 54685925 IUPAC Name: (4S,4aS,5aR,12aR)-4,7-bis(dimethylamino)-1,10,11,12a-tetrahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
Minocycline hydrochloride
CAS: 13614-98-7 Molecular Formula: C23H28ClN3O7 Molecular Weight (g/mol): 493.94 MDL Number: MFCD00083669 InChI Key: KDLQIOPKJDNQIM-WUURTAMISA-N PubChem CID: 54685925 IUPAC Name: hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
Minocycline Hydrochloride, MP Biomedicals™
CAS: 13614-98-7 Molecular Formula: C23H28ClN3O7 Molecular Weight (g/mol): 493.94 MDL Number: MFCD00083669 InChI Key: KDLQIOPKJDNQIM-WUURTAMISA-N Synonym: minocycline hydrochloride,minocycline hcl,minomycin,minocin,minocycline chloride,arestin,vectrin,minomax,periocline,tri-mino PubChem CID: 54685925 IUPAC Name: hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
Minocycline Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Minocycline Hydrochloride 98.0+%, TCI America™
CAS: 13614-98-7 Molecular Formula: C23H28ClN3O7 Molecular Weight (g/mol): 493.94 MDL Number: MFCD00083669 InChI Key: KDLQIOPKJDNQIM-WUURTAMISA-N Synonym: minocycline hydrochloride,minocycline hcl,minomycin,minocin,minocycline chloride,arestin,vectrin,minomax,periocline,tri-mino PubChem CID: 54685925 IUPAC Name: hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
R&D Systems™ Minocycline hydrochloride
CAS: 13614-98-7 Molecular Formula: C23H28ClN3O7
Minocycline HCl, USP Grade, G-Biosciences
CAS: 13614-98-7 Molecular Weight (g/mol): 493.94 g/mol