Learn More
Enzo Life Sciences Minocycline. hydrochloride (50mg). CAS: 13614-98-7
SDP
Supplier:  Enzo Life Sciences ALX380109M050
Encompass_Preferred
Tetracycline derivative with antimicrobial activity. Inhibitor of angiogenesis, apoptosis and poly(ADP-ribose) polymerase-1 (PARP-1). MMP inhibitor. Anti-inflammatory and neuroprotective. Alternative name: Dynacin, Klinomycin, Minocin, Vectrin. Purity: ≥98% (UHPLC). Appearance: Yellow crystalline solid. Solubility: Soluble in water (50mg/ml, warm) or DMSO (7mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8960
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.