Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Minocycline hydrochloride, 97%

Catalog No. AC455330010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455330010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455330010 Supplier Thermo Scientific Chemicals Supplier No. 455330010
  • $244.00
    $227.65 / Each of 1

    Save $16.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Minocycline hydrochloride, 97%, CAS # 13614-98-7, is a broad-spectrum compound with antibacterial and anti-inflammatory activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Minocycline hydrochloride is the hydrochloride salt of minocycline, a broad spectrum long-acting derivative of the antibiotic tetracycline, with antibacterial and anti-inflammatory activities

• This compound exerts its action by binding to the bacterial 30S ribosomal subunit and interfering with the binding of tRNA to the ribosomal complex, resulting in the inhibition of protein translation in bacteria

• Minocycline hydrochloride can inhibit the inflammatory enzyme 5-lipoxygenase (5LOX) and may block T cell-microglia interactions. 5LOX catalyzes the synthesis of inflammatory mediators, including prostaglandins and leukotrienes. Therefore, both molecular activities described may contribute to minocycline's neuroprotective activity.

Applications

• This compound is effective against tetracycline-resistant Staphylococcus cultures • Minocycline hydrochloride inhibits lipopolysaccharide mediated inflammatory cytokine tumor necrosis factor (TNF-α) secretion by macrophages

• It inhibits endothelial cell proliferation and angiogenesis in vitro through the translational suppression of HIF-1α

• This compound exhibits anti-tumor activity in glioma by inhibiting membrane type 1 matrix metalloproteinase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7
Molecular Weight (g/mol) 493.94
MDL Number MFCD00083669
InChI Key KDLQIOPKJDNQIM-WUURTAMISA-N
PubChem CID 54685925
IUPAC Name (4S,4aS,5aR,12aR)-4,7-bis(dimethylamino)-1,10,11,12a-tetrahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Type Minocycline Hydrochloride
Infrared Spectrum Authentic
Water 10% max.
Quantity 1 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in dmf and methanol
Formula Weight 493.94
Percent Purity 97%
Chemical Name or Material Minocycline hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.