Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Minocycline Hydrochloride, >98% Purity, Solid, 50 mg

Catalog No. 502707373
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7373 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7373 Supplier Abcam Supplier No. AB12066150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Antibiotic agent with anti-inflammatory, antioxidant, and neuroprotective properties that is blood-brain barrier permeable.

  • Molecular weight: 493.94 g/mol
  • Purity: >98%
  • Physical form: Solid
  • Function: Antibiotic
  • Biological activities: Demonstrates anti-inflammatory, antiapoptotic, antioxidant, and neuroprotective effects
  • Properties: Blood-brain barrier permeable
  • Inhibitor: Matrix metalloproteinase (MMP) inhibitor (IC50 = 290 μM at stromelysin (MMP-3))
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Matrix Metalloproteinase (MMP-3)
Shipping Condition Ambient
Buffer 100% Minocycline hydrochloride
Product Type Antioxidant
Content And Storage Store at 4°C.
Form Solid
Formulation 100% Minocycline hydrochloride
Quantity 50 mg
For Use With (Application) Antibiotic, Anti-inflammatory, Antiapoptotic, Antioxidant, Neuroprotective, Matrix metalloproteinase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.