Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minocycline Hydrochloride, MP Biomedicals™

Catalog No. ICN15571880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15571880 100 mg
ICN15571883 250 mg
ICN15571891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15571880 Supplier MP Biomedicals, Inc Supplier No. 0215571880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7
Molecular Weight (g/mol) 493.94
MDL Number MFCD00083669
InChI Key KDLQIOPKJDNQIM-WUURTAMISA-N
Synonym minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino
PubChem CID 54685925
IUPAC Name hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
SMILES [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 493.9
Physical Form Powder
Chemical Name or Material Minocycline Hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.