Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catalog Number | Mfr. No. | Model | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Size | Price | Quantity & Availability | ||||
10-001-559
![]() |
Stoelting™
5213332 |
Black | 0.15 mm |
Pack of 500 for $120.00
|
|
||||
10-001-560
![]() |
Stoelting™
5213333 |
Stainless steel | 0.15 mm |
Pack of 500 for $120.00
|
|
||||
10-001-561
![]() |
Stoelting™
5213334 |
Stainless steel | 0.1 mm |
Pack of 500 for $120.00
|
|
||||
10-001-562
![]() |
Stoelting™
5213335 |
Stainless steel | 0.2 mm |
Pack of 500 for $120.00
|
|
||||
Specifications
500 Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title