Learn More
MIOX Mouse anti-Human, Clone: OTI5F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501424

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MIOX (Myo-Inositol Oxygenase) is a Protein Coding gene. Among its related pathways are superpathway of inositol phosphate compounds and Porphyrin and chlorophyll metabolism. Gene Ontology (GO) annotations related to this gene include iron ion binding and oxidoreductase activity, acting on NAD(P)H.Specifications
MIOX | |
Monoclonal | |
Unconjugated | |
MIOX | |
ALDRL6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55586 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UGB7 | |
MIOX | |
Full length human recombit protein of human MIOX produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.