Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MIP-3 alpha/CCL20 Cell Lysate ELISA Kit

Catalog No. 502741178
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-1178 10 x 96 Tests
EHCCL20CL 96 Tests
EHCCL20CLX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-274-1178 Supplier Invitrogen™ Supplier No. EHCCL20CLX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Human MIP-3 alpha/CCL20 ELISA quantitates Hu MIP-3α in human cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Hu MIP-3α. Principle of the method The Human MIP-3α solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78556
Assay Range 1.5-600 pg/mL
Assay Sensitivity 1.5 pg/mL
Calibration Range 1.5 pg/mL to 600 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chemokines & Receptors
Sample Type Cell Lysates, Plasma, Serum, Supernatant, Tissue Homogenates
Specificity The antibody pair provided in this kit recognizes human MIP-3 alpha.
Sufficient For 10 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 α; MIP 3alpha; MIP 3α; MIP3 a; MIP3 alpha; MIP3 α; MIP-3[a]; MIP3A; MIP-3A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 α; MIP 3alpha; MIP 3α; MIP3 a; MIP3 alpha; MIP3 α; MIP-3[a]; MIP3A; MIP-3A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38; CKb4; Exodus; LARC; MIP-3-alpha; MIP-3a; MIP3A; SCYA20; ST38
Gene ID (Entrez) 6364
Gene Symbol CCL20
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Cell Lysis Buffer, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Cell Lysis Buffer, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Cell Lysate 20 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym CKb4, Exodus, LARC, MIP-3-alpha, MIP-3 A, MIP3A, SCYA20, ST38
Target CCL20
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.