Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MIRA-1, p53 Inducer, 99% Purity, Solid, 10 mg

Catalog No. 502705067
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5067 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5067 Supplier Abcam Supplier No. AB14586210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Novel, nontoxic, and selective antitumor agent that promotes tumor cell apoptosis in a mutant p53-dependent manner.

  • Purity: >99%
  • Molecular Weight: 183.16 Da
  • Mechanism: Novel, nontoxic and selective p53 inducer; promotes tumor cell apoptosis in a mutant p53-dependent manner (IC50 = 10 μM)
  • Application: Antitumor agent; active in vivo and in vitro
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule p53
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Content And Storage Store at 4°C.
Format Vial
Form Solid
Formulation 100% (2,5-dioxopyrrol-1-yl)methyl propanoate
Quantity 10 mg
For Use With (Application) Antitumor research, Apoptosis study
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.