Learn More
Apexbio Technology LLC Meprednisone 1247-42-3 5mg
Supplier: Apexbio Technology LLC B20815
Meprednisone (CAS 1247-42-3) is a small-molecule agonist targeting the glucocorticoid receptor (GR) It is designed to activate glucocorticoid receptors thereby modulating gene transcription involved in inflammatory and immune response pathways Meprednisone exerts its biological activity primarily through interaction with intracellular glucocorticoid receptors influencing the regulatory mechanisms underlying anti-inflammatory and immunosuppressive responses Based on these pharmacological properties Meprednisone holds research potential in the exploration of glucocorticoid-mediated anti-inflammatory and immunosuppressive mechanisms and as a reference compound in comparative receptor-binding and functional activity assays
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.