Learn More
Medchemexpress LLC Mirtazapine-d3 | 1216678-68-0 | MFCD09841051 | 98.6% | 268.38 | C17H16D3N3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0352S10MG
Encompass_Preferred
Mirtazapine-d3 is a deuterium-labeled analogue of mirtazapine used as an internal standard and tracer for quantitative analytical methods. It delivers a mass-shifted reference for accurate quantitation in pharmacokinetic, bioanalytical, and metabolite studies, improving precision in LC-MS, GC-MS, and NMR assays.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Compatible with LC-MS, GC-MS, and NMR techniques.
  • High purity for consistent analytical performance.
  • Powder form with defined cold-storage stability.
Catalog No. 50-002-97769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.