Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirtazapine | 85650-52-8 | MFCD00865427 | 265.36 | C17H19N3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0352R10MG

Encompass_Preferred

Mirtazapine analytical reference standard for research and analytical applications, provided as a small-mass powdered material suitable for preparing stock solutions and calibrants. It represents the standard form of mirtazapine used in method development, calibration, and quality control workflows.

  • Analytical reference suitable for method development and calibration.
  • Used in studies targeting 5-HT, histamine H1, and adrenergic receptors.
  • Molecular formula C17H19N3 and molecular weight 265.36 g/mol.
  • Available in small mass packs (for example, 1 mg, 5 mg, 10 mg) for stock preparation.
  • Storage guidance provided for powder and solutions to preserve stability.

Catalog No. 50-002-98190


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.