Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Amine Modifying Reagent

Catalog No. MIR3925 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
For 100 μg nucleic acid
For 25 μg nucleic acid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR3925 For 25 μg nucleic acid
MIR3900 For 100 μg nucleic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR3925 Supplier Mirus Bio™ Supplier No. MIR 3925
Add to Cart
Add to Cart

Attach amine functional groups covalently to nucleic acids. The simplicity of this chemical labeling reaction allows one to control the level of modification. For labeling with hydrazine, amine, or amine-reactive dyes and attachment to activated glass surfaces or hydrazide-coated beads or plates.

  • Modify any DNA or RNA Template: Direct, covalent attachment of amine functional groups to any nucleic acid
  • One-step chemical method: Easily and consistently control the density of nucleic acid modification
  • Covalent mechanism: Permanent, nondestructive modification of nucleic acids which can then be conjugated to proteins or peptides, or attached to glass surfaces, or beads or plates

Specifications

Detection Method Fluorescence
Label or Dye Not Applicable
Final Product Type DNA (Labeled), RNA (Labeled)
Labeling Target DNA (General), Oligos, RNA (General)
For Use With (Application) Labeling with Hydrazine, Amine or Amine-reactive dyes, attachment to activated Glass surfaces or Hydrazide-coated beads or plates
Includes Label IT™ Reagent, Label IT™ reconstitution solution, Labeling Buffer, solutions and G50 spin columns
Product Type Modifying Reagent
Quantity For 25 μg nucleic acid
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.