Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ siRNA Tracker™ Intracellular Localization Kit

Catalog No. MIR7216 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Product Type:
Cy3 Labeling Kit
Fluorescein Labeling Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
MIR-7216 Fluorescein Labeling Kit
MIR-7212 Cy3 Labeling Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR-7216 Supplier Mirus Bio™ Supplier No. MIR7216

missing translation for 'validMainframeMsg'

Includes

Label IT siRNA Tracker Reagent, Label IT Reconstitution Solution, and 10X Labeling Buffer A

Label, transfect, and track any siRNA residues in an efficient and nondestructive manner. LabelIT siRNA Tracker Intracellular Localization Kit provides a straight-forward approach to directly label siRNA of any sequence containing guanine residues.

  • Generate a labeled siRNA sample without hindering its inherent ability to silence targeted genes.
  • Monitor both sub-cellular localization and functional inhibition of target gene expression simultaneously following introduction of the labeled siRNA into mammalian cells
  • Contain sufficient reagents to label 50μg of siRNA
  • siRNA can be delivered to cells in vitro or in vivo using the transfection reagent of your choice

Specifications

Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Vial
Promoter None (Promoterless)
Transfection Technique Lipid-based Transfection, Transient Transfection
For Use With (Application) Monitor both subcellular localization and functional inhibition of target gene expression simultaneously following introduction of the labeled siRNA into mammalian cells
Includes Label IT™ siRNA tracker reagent, reconstitution solution, 10X Labeling Buffer A and siRNA dilution Buffer
Product Type Fluorescein Labeling Kit
Quantity 1 Ea.
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.