Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-2020 Reagent

Catalog No. MIR5400 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 x 0.4 mL
1 x 1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR5400 1 x 1 mL
MIR5404 1 x 0.4 mL
MIR5405 5 x 1 mL
MIR5406 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. MIR5400 Supplier Mirus Bio™ Supplier No. MIR5400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

A high performance, animal-free, broad spectrum DNA transfection reagent for delivery of plasmid DNA into mammalian cells. Suitable for both transient and stable transfection. Reagent does not require medium changes and can be performed in serum-containing medium.

A high performance, animal-free, broad spectrum DNA transfection reagent for delivery of plasmid DNA into mammalian cells. Suitable for both transient and stable transfection. Reagent does not require medium changes and can be performed in serum-containing medium.

  • Broad Spectrum DNA Delivery: Achieve high expression in many cell types, including hard-to-transfect cell lines and primary cells
  • Outperforms Competitor Reagents: TransIT-2020 demonstrated higher protein yield when compared to Lipofectamine™2000 and Lipofectamine™LTX and PLUS™
  • Animal Origin Free: TransIT-2020 provides high performance with maximum compatibility
High efficiency transfection of DNA suitable for hard-to-transfect cells including primary and stem cells

Specifications

Content And Storage -20°C
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Provides exceptional transfection of plasmid DNA into mammalian cells
Product Type Transfection Reagent
Quantity 1 x 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.