Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-293 Transfection Reagent
GSA_VA

Catalog No. MIR2706 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 x 0.4 mL
1 x 1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2706 10 x 1 mL
MIR2704 1 x 0.4 mL
MIR2700 1 x 1 mL
MIR2705 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2706 Supplier Mirus Bio™ Supplier No. MIR 2706
Add to Cart
Add to Cart
Request a Quote

TransIT™-293 Transfection Reagent designed for high-efficiency, low-toxicity DNA transfection in HEK293 cells.

  • Optimizes transfection performance in HEK 293 cells
  • Low cellular toxicity maintains cell density and reduces experimental bias
  • Allows 75 to 85% transfection efficiency in HEK 293 cell lines
  • Delivers single or multiple plasmid for specialized applications such as virus production
  • Supplied as a ready-to-use solution
  • Concentration: 1.41 mg/mL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Store for up to one year at 4°C (do not freeze).
Cell Type 293-derived Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 1.41 mg/mL
For Use With (Application) High efficiency, low-toxicity DNA transfection in HEK293 cells
Product Type Transfection Reagent
Quantity 10 x 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.