Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-BrCa Transfection Reagent

Catalog No. MIR5506 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR5506 10 x 1 mL
MIR5504 0.4 mL
MIR5500 1 mL
MIR5505 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR5506 Supplier Mirus Bio™ Supplier No. MIR5506
Add to Cart
Add to Cart

Dedicated breast-cancer-cell-line-specific reagent. Delivers efficiently to many breast cancer cell types and maintains low cellular toxicity. Transfection with TransIT™-BrCa Reagent does not require medium changes and can be performed in serum-containing medium.

  • Broad spectrum DNA delivery: Achieve high expression levels in breast cancer cell types including MCF-7, MDA-MB-231, MDA-MB-453, MDA-MB-468 and T47D
  • Formulated for low cellular toxicity: Maintain cell density and reduce experimental biases due to toxicity
  • Multiple applications: Transient and stable transfections for gene expression, shRNA expression and promoter analyses

Specifications

Cell Type Cancer Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Achieve high expression levels in breast cancer cell types including MCF-7, MDA-MB-231, MDA-MB-453, MDA-MB-468 and T47D, transient and stable transfections for gene expression, shRNA expression and promoter analyses
Product Type Transfection Reagent
Quantity 10 x 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.