Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-CHO Transfection Kit

Catalog No. MIR2175 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2175 5 mL
MIR-2170 1 mL
MIR-2174 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MIR2175 Supplier Mirus Bio™ Supplier No. MIR 2175

missing translation for 'validMainframeMsg'

Includes

TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.

TransIT™-CHO Transfection Kit developed for high transfection efficiency and cell viability in CHO cell types. Does not require medium changes and can be performed in serum-containing medium.

  • TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.
  • Unique Formulation: Each kit contains a base reagent and a booster reagent to optimize transfection performance in CHO cells
  • Low Cellular Toxicity: Maintains cell density and reduces experimental bias
  • High Efficiency Delivery: Allows 50 to 60% transfection efficiency in CHO-K1 cells
  • Suitable for both transient and stable transfection

Specifications

Content And Storage Store at -20°C
Cell Type CHO
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Specifically optimized to provide exceptional transfection efficiency of plasmid DNA in CHO cells and cell types of associated lineage
Includes 5 mL 2.22 mg/mL in 80% Ethanol, TransIT™-CHO reagent and 3.5 mL 5 mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution
Product Type Transfection Reagent
Quantity 5 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.