Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Jukart Transfection Reagent

Catalog No. MIR2126 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
5 x 1 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR-2126 10 x 1 mL
MIR-2125 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR-2126 Supplier Mirus Bio™ Supplier No. MIR 2126
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution

TransIT™-Jukart Transfection Reagent optimized to achieve 10 – 25% transfection efficiency in Jurkat cells.

  • TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution.
  • Cells can be transfected in serum-containing media, eliminating the need for media changes and improving transfection effeciency.
  • Concentration: 10 mg/mL

Specifications

Cell Type Primary Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 10 mg/mL
For Use With (Application) Gene expression and promoter analysis, optimized for Jurkat cells and other hard-to-transfect cells
Product Type Transfection Reagent
Quantity 10 x 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.