Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-mRNA Transfection Kit

Catalog No. MIR2256 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2256 10 mL
MIR-2255 5 mL
MIR-2250 1 mL
MIR-2225 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2256 Supplier Mirus Bio™ Supplier No. MIR2256
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

TransIT-mRNA Transfection Reagent; mRNA Boost Reagent

Developed specifically for high efficiency delivery of both mRNA and viral RNA to various types of mammalian cells. Optimized for use in presence of serum, saving assay time and eliminating media changes. Transcriptional regulation effects are avoided by directly delivering the RNA to the cytoplasm.

  • Low Cellular Toxicity: Maintain cell density and reduce experimental biases
  • High Efficiency Delivery: Achieve RNA delivery in large population of cells to ensure experimental success
  • Serum Compatible: Perform transfections in presence of serum which eliminates need for media change and maintains cellular health
  • Deliver Various Sizes of RNA: Ideal for specialized applications, such as viral production and protein expression from mRNA
High efficiency delivery of both mRNA and viral RNA

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) High efficiency delivery of both mRNA and viral RNA to various types of mammalian cells
Includes 10 mL TransIT™-mRNA Transfection Reagent, 10 mL mRNA Boost Reagent
Product Type Transfection Reagent
Quantity 10 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.