Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Oligo Transfection Reagent

Catalog No. MIR2160 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR-2160 1 mL
MIR-2165 5 x 1 mL
MIR-2166 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MIR-2160 Supplier Mirus Bio™ Supplier No. MIR 2160
Add to Cart
Add to Cart

TransIT™-Oligo Transfection Reagent provides efficient transfection of a wide range of oligonucleotides and oligoribonucleotides into a variety of cell types. Transfections do not require media changes and can be performed in serum-containing media.

  • Unique Formulation: Maximize transfection performance of oligonucleotides into a wide range of cells
  • Low Cellular Toxicity: Maintain cell density and reduce experimental biases
  • High Efficiency Delivery: Achieve high transfection efficiency in cells to ensure experimental success
  • One milliliter is sufficient to perform up to 500 transfections in 24-well plates
  • Tested on a variety of cell lines and oligonucleotides

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Provides efficient transfection of a wide range of Oligonucleotides and Oligoribonucleotides into a variety of cell types
Product Type Transfection Reagent
Quantity 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.