Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-PRO™ Transfection Kit

Catalog No. MIR5700 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR5700 1 mL
MIR5760 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR5700 Supplier Mirus Bio™ Supplier No. MIR5700

missing translation for 'validMainframeMsg'

Requires

Protein production cell line, culture vessels, appropriate cell culture medium, purified DNA, serum-free medium, sterile tube for transfection complex preparation, micropipets, orbital shaker, reporter assay (as required).

Specifically developed for mammalian protein production in suspension CHO and 293 cells. The PRO Boost Reagent is optional and enhances expression in certain media formulations. Use of TransIT-PRO Transfection Kit eliminates the need for a culture medium change post-transfection.

  • Suitable for both transient and stable transfection
  • Compatible with multiple CHO media formulations
  • Reproducible protein expression with minimal optimization
  • Outperforms linear PEI in protein yield and provides a cost-effective alternative to FreeStyle™ MAX

Warranty and Services

One year (when properly stored and handled).

Specifications

Content And Storage -20°C
Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Mammalian Protein production in suspension 293 and CHO derived cells, eliminates the need for a culture medium change post-transfection and is suitable for both transient and stable transfection
Includes 1 mL TransIT-PRO™ Reagent, 1.5 mL PRO™ Boost Reagent
Product Type Transfection Reagent
Quantity 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.