Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-QR Hydrodynamic Delivery Solution and Starter Kit

Catalog No. MIR5240 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
40 Injections
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR-5240 40 Injections
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR-5240 Supplier Mirus Bio™ Supplier No. MIR 5240
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TransIT™-QR Hydrodynamic Delivery Solution and Starter Kit offers a nontoxic, Quick Recovery in vivo delivery solution that delivers DNA or siRNA to the livers of laboratory mice using methods including hydrodynamic tail vein injection.

  • Naked Nucleic Acid Delivery: Achieve higher levels of gene expression than compared to polylysine and PEI methods
  • Versatile Platform: For DNA or siRNA delivery using methods including hydrodynamic tail vein injection to the mouse strain of your choice
  • Low Toxicity: Minimized loss of cardiac output compared to saline injections, which allows for quick recovery (within minutes) of the mouse post-injection
  • Concentration: 1X

Specifications

Cell Type Cells in Vivo
Format Bottle
Promoter None (Promoterless)
Concentration 1X
For Use With (Application) Delivers DNA or siRNA to the livers of laboratory mice using methods including hydrodynamic tail vein injection
Includes TransIT-QR Delivery Solution, 10 syringes, needles, alcohol swabs, and a conical plastic tube.
Product Type In vivo delivery solution
Quantity 40 Injections
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.