Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-TKO™ siRNA Transfection Reagent

Catalog No. MIR2156 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.4 mL
5 x 1.5 mL
10 x 1.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2156 10 x 1.5 mL
MIR2154 0.4 mL
MIR2155 5 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MIR2156 Supplier Mirus Bio™ Supplier No. MIR2156
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Enables high efficiency siRNA delivery and knockdown of target gene expression in many cell types, including primary cells. siRNA and DNA co-transfection is feasible with TransIT-TKO. Transfections do not require medium changes and can be performed in serum-containing medium.

  • Broad Spectrum siRNA Delivery: Utilize one transfection reagent and protocol for a wide variety of cells
  • Low Cellular Toxicity: Maintain cell density and reduce experimental biases
  • Reproducible Results: Obtain consistent, targeted gene knockdowns from day to day
  • High Knockdown Efficiency: Achieve optimal gene silencing in a large percentage of cells to ensure experimental success
  • Concentration: 1.37 mg/mL

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 1.37 mg/mL
For Use With (Application) Transfect small interfering RNA for knock out of target gene expression, optimal gene silencing in a large percentage of cells
Product Type Transfection Reagent
Quantity 10 x 1.5 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.