Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ MiraCLEAN™ Endotoxin Removal Kit

Catalog No. MIR5910 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
For 10 mg DNA
For 100 mg DNA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR5910 For 10 mg DNA
MIR5900 For 100 mg DNA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR5910 Supplier Mirus Bio™ Supplier No. MIR 5910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses rapid phase extraction to remove bacterial endotoxins from nucleic acids for both in vivo and in vitro applications. The proprietary pink-colored EndoGO extraction reagent helps the user to visualize the interface between phases, facilitating phase separation and increasing nucleic acid recovery.

  • MIR 5910 provides sufficient reagents for three endotoxin extraction rounds on a total of at least 10 mg of DNA
  • MIR 5900 provides sufficient reagents for three endotoxin extraction rounds on a total of at least 100 mg of DNA
  • Effectively Removes Endotoxin from Plasmid Samples: Aids in the safety and efficiency of gene delivery research
  • Compatible with In Vivo and In Vitro Applications: Removes harmful endotoxin that could cause decreased transfection efficiencies in vitro and inflammatory reactions in vivo
  • Ease of Use Simple phase separation protocol allows quick visualization of recovered nucleic acid preparations

Specifications

Content And Storage 4°C
Format Kit
Isolation Technology Organic Extraction
For Use With (Application) Removes bacterial endotoxins from Nucleic Acids for both in vivo and in-vitro applications
Product Type Endotoxin Removal Kit
Quantity For 10 mg DNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.