Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ pLIVE™ In Vivo Expression and Reporter Vectors

Catalog No. MIR5320 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR5320 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR5320 Supplier Mirus Bio™ Supplier No. MIR 5320
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pLIVE™ In Vivo Vector Complete System is a vector system designed to achieve long term expression in the livers of laboratory mice.

  • Sustained Gene Expression: Achieve long-term gene expression (up to 8 months) in the liver post hydrodynamic tail vein injection
  • Versatile Platform: Available with positive control vectors expressing either LacZ or SEAP
  • pLIVE Vector Complete System includes 20 μg each of pLIVE vector, pLIVE Vector/LacZ Control Vector, pLIVE Vector/SEAP Control Vector
  • pLIVE Vector /Lac Z Control Vector Kit includes 20 μg each of pLIVE Vector and pLIVE/Lac Z Control Vector

Specifications

Includes pLIVE Vector Complete System: 20 μg each of pLIVE vector, pLIVE Vector/LacZ Control Vector, pLIVE Vector/SEAP Control Vector, pLIVE Vector /Lac Z Control Vector Kit: 20 μg each of pLIVE Vector, pLIVE/Lac Z Control Vector
Product Type Expression Reporter Vector
Storage Buffer TE Buffer (10mMTris, pH 7.5, 0.1mM EDTA)
Content And Storage -20°C
Cloning Method Restriction Enzyme/MCS
Concentration 1 mg/mL
Primer Custom Primer
Quantity 20 μg
Vector 3423 bp pLIVE vector, 6509 bp pLIVE lacZ vector, 4992 bp pLIVE-SEAP vector
Product Line pLIVE
For Use With (Application) Long term expression in the livers of laboratory Mice
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.