Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Selection Antibiotic: G418 Sulphate Solution

Catalog No. MIR5920 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Sterilization Method:
0.2 μm Sterile-filtered
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterilization Method
MIR5920 0.2 μm Sterile-filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR5920 Supplier Mirus Bio™ Supplier No. MIR5920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for positive selection of cells that express the neogene encoding neomycin phosphotransferase

Different cell types and cell culture conditions may require different concentrations of selection antibiotic. Perform a kill curve to determine the optimal working concentration for your experiment.

  • Recommended concentration for selection of mammalian cells resistant to G418 is 100 to 2000 μg/mL
  • 10 mL solution (50 mg/mL in DI water
  • Sterile filtered

Warranty and Services

1 year

Specifications

Chemical Name or Material G418 Sulphate Solution
Classification Antibiotic
Concentration 100 to 2000 μg/mL
Culture Type Cell Culture
Form Solution
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
For Use With (Application) Stable cell line generation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.