Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-LT1 Transfection Reagent
GSA_VA

Catalog No. mir2305 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2305 5 x 1 mL
MIR2304 0.4 mL
MIR2300 1 mL
MIR2306 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2305 Supplier Mirus Bio™ Supplier No. MIR 2305
Only null left
Add to Cart
Add to Cart

High-efficiency plasmid DNA delivery in many mammalian cell types, including primary cells. The TransIT™-LT1 Transfection Reagent is broad spectrum, low toxicity, and serum compatible, and is suitable for both transient and stable transfection.

  • Broad spectrum DNA delivery: utilize one transfection reagent and protocol for a variety of cells
  • Low cellular toxicity: maintain cell density and reduce experimental biases
  • High efficiency delivery: achieve expression in a large population of cells for experimental success
  • Deliver single or multiple plasmids: suitable for many applications such as gene expression, shRNA expression, viral production, and promoter analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Store for up to one year at 4°C.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Vial
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Gene expression, shRNA expression, viral production and promoter analysis
Product Type Transfection Reagent
Quantity 5 x 1 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.