Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mis12 Complex Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE258525UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2585-25UG 25 μg
ABE2585MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE2585-25UG Supplier MilliporeSigma Supplier No. ABE258525UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Mis12 Complex Antibody is a highly specific rabbit polyclonal antibody that targets Mis12 complex and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mis12 Complex
Applications Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols MIS12,PMF1,DSN1,NSL1
Host Species Rabbit
Immunogen Four subunits from human Mis12 complex.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This polyclonal antibody detects Mis12 complex in human. It will also detect individual subunits (Mis12, PMF1, Dsn1, Nsl1) in Western Blotting.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.