Learn More
Cayman Chemical Misoprostol, 59122-46-2, 10 mg
SDP
Supplier:  Cayman Chemical 1382010
Encompass_Preferred
Misoprostol is a PGE1 analog with agonist activity mediated by EP2, EP3, and EP4 receptors.{2313,1754,1752,1611} It has been shown to inhibit the formation of gastric lesions in rats (ED50 = 0.31 µg/kg),{1754} inhibit superoxide generation in human neutrophils (EC50 = 0.35 µM),{1752} and relax fetal rabbit ductus arteriosus (EC50 = 0.36 nM){1611} in a concentration dependent manner. Misoprostol is commonly used in clinical medicine for the prevention of peptic ulcer disease. It has also been used in conjunction with RU-486 for the oral induction of first trimester abortion. Misoprostol is readily absorbed and rapidly hydrolyzed in humans to the active free acid.{2313}
Catalog No. NC0815650
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.