Learn More
Abcam Misoprostol, Cytoprotective PGE2 Analog, Liquid, >98% Purity, 1 mg
Description
- Molecular weight: 382.54 g/mol
- Purity: >98%
- Description: Cytoprotective PGE2 analog
- Activity: Mediated by EP2, EP3, and EP4 receptors (Ki values: 250, 67, and 67 nM, respectively)
- Form: Liquid
- Storage buffer: 99% Methyl acetate, 1% Misoprostol
- Storage conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 99% methyl acetate, 1% misoprostol |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Blue Ice |
| Target Molecule | Prostaglandin E2 (EP2, EP3, and EP4 receptors) |
| Form | Liquid |
| Formulation | 99% Methyl acetate, 1% Misoprostol |
| Quantity | 1 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.